Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl glutamate | 1119-33-1 | MFCD00037217 | 97.0% | 175.18 g·mol⁻1 | C7H13NO4 | 25G
SDP

Catalog No. 5000238627
Encompass_Preferred

Ethyl glutamate (L-glutamic acid γ-ethyl ester; CAS 1119-33-1) is an amino-acid derivative used as an endogenous metabolite reagent in life-science research. It is provided as a 25 g research reagent for studies of glutamate metabolism and cell growth.

  • Molecular formula: C7H13NO4.
  • Molecular weight: 175.18 g·mol⁻1.
  • Purity: 97.0%.
  • Available pack: 25 g solid.
  • Typical applications: reagent for biochemical assays and cell-biology studies.
  • Intended use: for research use only, not for diagnostic or therapeutic applications.

Catalog No. 50-002-38627 Supplier Medchemexpress LLC Supplier No. HY12422825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl glutamate (L-glutamic acid γ-ethyl ester; CAS 1119-33-1) is an amino-acid derivative used as an endogenous metabolite reagent in life-science research. It is provided as a 25 g research reagent for studies of glutamate metabolism and cell growth.

  • Molecular formula: C7H13NO4.
  • Molecular weight: 175.18 g·mol⁻1.
  • Purity: 97.0%.
  • Available pack: 25 g solid.
  • Typical applications: reagent for biochemical assays and cell-biology studies.
  • Intended use: for research use only, not for diagnostic or therapeutic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.