Learn More
Medchemexpress LLC Ethyl glutamate | 1119-33-1 | MFCD00037217 | 97.0% | 175.18 g/mol | C7H13NO4 | 100G
SDP
Catalog No. 5000238448
Encompass_Preferred
Ethyl glutamate is an amino-acid derivative (the ethyl ester of glutamic acid) used as an endogenous metabolite and research reagent in life-science studies. It is reported to promote cell growth and is supplied as a white to off-white solid with molecular weight 175.18 g/mol and formula C7H13NO4. Typical uses include biochemical assays, metabolite profiling, and cell-culture research; handle according to the safety data sheet.
Catalog No. 50-002-38448 Supplier Medchemexpress LLC Supplier No. HY124228100G
Ethyl glutamate is an amino-acid derivative (the ethyl ester of glutamic acid) used as an endogenous metabolite and research reagent in life-science studies. It is reported to promote cell growth and is supplied as a white to off-white solid with molecular weight 175.18 g/mol and formula C7H13NO4. Typical uses include biochemical assays, metabolite profiling, and cell-culture research; handle according to the safety data sheet.