Learn More
Description
- Ethyl glycollate is an ester. microwave spectra of ethyl glycollate and its deuterated species has been reported.ethyl glycollate is an important building block for the preparation of medicinal compounds and agrochemicals. it may be used:
- As starting reagent in the synthesis of 2-ferrocenylfuran
- As substrate during the polycondensation of α-hydroxy acids by enzymes and polyethylene glycol (peg)-modified enzymes
- In the synthesis of poly(pdl-ga) copolymers, via enzymatic catalysis
Specifications
Specifications
| CAS | 623-50-7 |
| Density | 1.1 g/mL (at 25°C (literature)) |
| Boiling Point | 158°C to 159°C (lit.) |
| Molecular Formula | C4H8O3 |
| Refractive Index | n20/D 1.419 (literature) |
| Linear Formula | HOCH2CO2C2H5 |
| MDL Number | MFCD00021970 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 104.1 |
| Percent Purity | 98% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.