Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl glycolate

Catalog No. 3648435G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-484-35G 5 g
36-484-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-484-35G Supplier Sigma Aldrich Supplier No. 3648435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl glycollate is an ester. microwave spectra of ethyl glycollate and its deuterated species has been reported.ethyl glycollate is an important building block for the preparation of medicinal compounds and agrochemicals. it may be used:
  • As starting reagent in the synthesis of 2-ferrocenylfuran
  • As substrate during the polycondensation of α-hydroxy acids by enzymes and polyethylene glycol (peg)-modified enzymes
  • In the synthesis of poly(pdl-ga) copolymers, via enzymatic catalysis

Specifications

CAS 623-50-7
Density 1.1 g/mL (at 25°C (literature))
Boiling Point 158°C to 159°C (lit.)
Molecular Formula C4H8O3
Refractive Index n20/D 1.419 (literature)
Linear Formula HOCH2CO2C2H5
MDL Number MFCD00021970
Quantity 5 g
Molecular Weight (g/mol) 104.1
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.