Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl heptanoate

Catalog No. 112364500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-236-4500ML 500 mL
11-236-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-236-4500ML Supplier Sigma Aldrich Supplier No. 112364500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl heptanoate

Specifications

CAS 106-30-9
Melting Point -66°C (lit.)
Density 0.87 g/mL (at 25°C (literature))
Boiling Point 188°C to 189°C (lit.)
Molecular Formula C9H18O2
Refractive Index n20/D 1.412 (literature)
Linear Formula CH3(CH2)5 COOC2H5
MDL Number MFCD00009538
Quantity 500 mL
Synonym Ethyl enanthate
Molecular Weight (g/mol) 158.24
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.