Learn More
Medchemexpress LLC Ethyl heptanoate | 106-30-9 | 99.7% | 158.24 | 100 G
SDP
Catalog No. 5000453589
Encompass_Preferred
Ethyl heptanoate is commonly used as a fragrance ingredient in various products, including food, beverages, and personal care items. It also serves as a solvent and a building block for synthesizing organic compounds like pharmaceuticals and agrochemicals. Due to its low toxicity and biodegradability, it has been explored for its potential as a bio-based solvent and for its antimicrobial properties against certain bacteria and fungi.
  • Commonly used as a fragrance ingredient.
  • Serves as a solvent and a building block for synthesizing organic compounds.
  • Explored for potential as a bio-based solvent.
  • Possesses antimicrobial properties against certain bacteria and fungi.
  • Exhibits low toxicity and biodegradability.
Catalog No. 50-004-53589 Supplier Medchemexpress LLC Supplier No. HYW010177100G
Ethyl heptanoate is commonly used as a fragrance ingredient in various products, including food, beverages, and personal care items. It also serves as a solvent and a building block for synthesizing organic compounds like pharmaceuticals and agrochemicals. Due to its low toxicity and biodegradability, it has been explored for its potential as a bio-based solvent and for its antimicrobial properties against certain bacteria and fungi.
  • Commonly used as a fragrance ingredient.
  • Serves as a solvent and a building block for synthesizing organic compounds.
  • Explored for potential as a bio-based solvent.
  • Possesses antimicrobial properties against certain bacteria and fungi.
  • Exhibits low toxicity and biodegradability.