Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl heptanoate | 106-30-9 | 99.74% | 158.24 | 500 G
SDP

Catalog No. 5000366387
Encompass_Preferred

Ethyl heptanoate is commonly used as a fragrance ingredient in a variety of products, including food, beverages, and personal care products. It can also be used as a solvent and a building block for the synthesis of various organic compounds, including pharmaceuticals and agrochemicals. Due to its low toxicity and biodegradability, it has been investigated for its potential use as a bio-based solvent, as well as for its potential antimicrobial properties against certain bacteria and fungi. It is a biochemical reagent suitable for biological material or organic compound research in life sciences.

  • Purity: 99.74%
  • CAS number: 106-30-9
  • Molecular weight: 158.24
  • Formula: C9H18O2
  • Appearance: Liquid (Density: 0.9±0.1 g/cm3)
  • Color: Colorless to light yellow

Catalog No. 50-003-66387 Supplier Medchemexpress LLC Supplier No. HYW010177500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl heptanoate is commonly used as a fragrance ingredient in a variety of products, including food, beverages, and personal care products. It can also be used as a solvent and a building block for the synthesis of various organic compounds, including pharmaceuticals and agrochemicals. Due to its low toxicity and biodegradability, it has been investigated for its potential use as a bio-based solvent, as well as for its potential antimicrobial properties against certain bacteria and fungi. It is a biochemical reagent suitable for biological material or organic compound research in life sciences.

  • Purity: 99.74%
  • CAS number: 106-30-9
  • Molecular weight: 158.24
  • Formula: C9H18O2
  • Appearance: Liquid (Density: 0.9±0.1 g/cm3)
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.