Learn More
Medchemexpress LLC Ethyl hexadecanoate-d31 | 1215721-57-5 | MFCD12022201 | 98.0% | 315.67 g/mol | C18H5D31O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN2086S10MG
Encompass_Preferred
Ethyl palmitate-d31 is the deuterium-labeled analogue of ethyl palmitate, provided as a stable-isotope reagent for analytical and research applications. It is a fatty acid ethyl ester used primarily in mass spectrometry and lipidomics to enable accurate quantitation and tracing of lipid species.
  • Deuterium-labeled stable isotope standard for analytical workflows.
  • Suitable as an internal standard in mass spectrometry and lipidomics.
  • High isotopic enrichment and 98.0% purity.
  • Useful for quantitation and tracer studies in lipid research.
  • Available in a 10 mg pack for small-scale laboratory use.
Catalog No. 50-002-96661