Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl hexanoate, 99%

Catalog No. AAA1398522
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1398522 100 g
AAA1398536 500 g
AAA139850E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1398522 Supplier Thermo Scientific Chemicals Supplier No. A1398522
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 123-66-0 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-66-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009511
InChI Key SHZIWNPUGXLXDT-UHFFFAOYSA-N
Synonym ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
PubChem CID 31265
ChEBI CHEBI:86055
IUPAC Name ethyl hexanoate
SMILES CCCCCC(=O)OCC

Specifications

Melting Point -67°C
Density 0.873
Boiling Point 166°C
Flash Point 49°C (121°F)
Refractive Index 1.4075
Quantity 100 g
UN Number UN3272
Merck Index 14,3777
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material Ethyl hexanoate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 204-640-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.