Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Hydrazine Oxalate, MP Biomedicals

Catalog No. ICN224575 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224575 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224575 Supplier MP Biomedicals, Inc Supplier No. 0522457505

Chemical Identifiers

CAS 6629-60-3
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
InChI Key DUMHBFMURBWDPC-UHFFFAOYSA-N
Synonym ethylhydrazine oxalate, ethyl hydrazine oxalate, ethylhydrazine, monooxalate, ethylhydrazine; oxalic acid, ethylhydrazine oxalate 1:1, hydrazine, ethyl-, ethanedioate 1:1, ethylhydrazinediylium oxalate, hydrazine, ethyl-, oxalate 1:1, ccris 7189, ethylhydrazine ethanedioate 1:1
PubChem CID 96181
IUPAC Name ethylhydrazine;oxalic acid
SMILES CCNN.C(=O)(C(=O)O)O

Specifications

Quantity 10 g
Formula Weight 150.14
Chemical Name or Material Ethyl Hydrazine Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.