Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl (hydroxyimino)cyanoacetate

Catalog No. 23341250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-341-250G 50 g
50-290-7049 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-341-250G Supplier Sigma Aldrich Supplier No. 23341250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl (hydroxyimino)cyanoacetate

Specifications

CAS 3849-21-6
Melting Point 130°C to 132°C (lit.)
Molecular Formula C5H6N2O3
Linear Formula NCC(=NOH)CO2C2H5
MDL Number MFCD00002112
Quantity 50 g
Synonym Ethyl cyano(hydroxyimino)acetate; Ethyl cyanoglyoxalate-2-oxime; Ethyl isonitrosocyanoacetate
Molecular Weight (g/mol) 142.11
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.