Learn More
Medchemexpress LLC Ethyl icosanoate (Icosanoic Acid Ethyl Ester) | 18281-05-5 | 99.6% | 340.59 | 5 G
SDP
Catalog No. 5000454262
Encompass_Preferred
Ethyl icosanoate is an ester formed by the esterification of eicosanoic acid and ethanol. It has a fruity, waxy smell and is used as a flavoring in foods, a fragrance ingredient in personal care products, and as a lubricant or plasticizer in industrial applications. It can also serve as a starting material for synthesizing other organic compounds.
  • Used as a flavoring in foods such as baked goods, candy, and beverages.
  • Utilized as a fragrance ingredient in personal care products.
  • Employed as a lubricant or plasticizer in various industrial applications.
  • Can be used as a starting material for the synthesis of other organic compounds.
  • Acts as a biochemical reagent for life science-related research.
Catalog No. 50-004-54262 Supplier Medchemexpress LLC Supplier No. HYW0132035G
Ethyl icosanoate is an ester formed by the esterification of eicosanoic acid and ethanol. It has a fruity, waxy smell and is used as a flavoring in foods, a fragrance ingredient in personal care products, and as a lubricant or plasticizer in industrial applications. It can also serve as a starting material for synthesizing other organic compounds.
  • Used as a flavoring in foods such as baked goods, candy, and beverages.
  • Utilized as a fragrance ingredient in personal care products.
  • Employed as a lubricant or plasticizer in various industrial applications.
  • Can be used as a starting material for the synthesis of other organic compounds.
  • Acts as a biochemical reagent for life science-related research.