Learn More
Medchemexpress LLC Ethyl indole-2-carboxylate | 3770-50-1 | 99.7% | 189.21 | 50 G
SDP
Catalog No. 5000403732
Encompass_Preferred
2-Carbethoxyindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • For research use only. We do not sell to patients.
  • An indole derivative.
  • Inhibits benzodiazepine receptor with IC50= 2.2 mM.
Catalog No. 50-004-03732 Supplier Medchemexpress LLC Supplier No. HY3469650G
2-Carbethoxyindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • For research use only. We do not sell to patients.
  • An indole derivative.
  • Inhibits benzodiazepine receptor with IC50= 2.2 mM.