Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Indoline-2-carboxylate 95.0+%, TCI America™
SDP

Catalog No. I05955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05955G Supplier TCI America Supplier No. I05955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50501-07-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD06797114
InChI Key KISPUTPAKVZNBI-UHFFFAOYSA-N
Synonym Indoline-2-carboxylic Acid Ethyl Ester
PubChem CID 11949691
IUPAC Name ethyl 2,3-dihydro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1CC2=CC=CC=C2N1

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Indoline-2-carboxylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.