Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals

Catalog No. AC222190100
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222190100 10 mL Glass bottle
AC222190500 50 mL Glass bottle
AC222192500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222190100 Supplier Thermo Scientific Chemicals Supplier No. 222190100
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-48-3
Molecular Formula C4H7IO2
Molecular Weight (g/mol) 214.00
MDL Number MFCD00001081
InChI Key MFFXVVHUKRKXCI-UHFFFAOYSA-N
Synonym ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
PubChem CID 12183
IUPAC Name ethyl 2-iodoacetate
SMILES CCOC(=O)CI

Specifications

Color Orange to Yellow
Density 1.8000g/mL
Boiling Point 179°C to 180°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5035 to 1.5055
Linear Formula ICH2CO2C2H5
Quantity 10 mL
Beilstein 02,222
Specific Gravity 1.8
Solubility Information Solubility in water: immiscible
Stabilizer copper
Formula Weight 214
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl iodoacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-796-3
RTECSNumber AI3575000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.