Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl iodoacetate, 98%, Thermo Scientific Chemicals

Catalog No. AAB2417714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2417714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2417714 Supplier Thermo Scientific Chemicals Supplier No. B2417714
  • $113.00
    $96.65 / Each of 1

    Save $16.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark. Storage temperature is 2-8°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-48-3
Molecular Formula C4H7IO2
Molecular Weight (g/mol) 214.00
MDL Number MFCD00001081
InChI Key MFFXVVHUKRKXCI-UHFFFAOYSA-N
Synonym ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
PubChem CID 12183
IUPAC Name ethyl 2-iodoacetate
SMILES CCOC(=O)CI

Specifications

Density 1.81
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Refractive Index 1.503
Quantity 25 g
UN Number UN2927
Beilstein 741934
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 214
Percent Purity 98%
Chemical Name or Material Ethyl iodoacetate
Show More Show Less
Hazard Category H227-H300-H311+H331-H314-H335
Hazard Statement GHS H Statement
H300-H314-H318-H227
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 210-796-3
RTECSNumber AI3575000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.