Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Isobutyrylacetate 99%, Thermo Scientific™

Catalog No. AC118610100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118610100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC118610100 Supplier Thermo Scientific Chemicals Supplier No. 118610100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7152-15-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
InChI Key XCLDSQRVMMXWMS-UHFFFAOYSA-N
Synonym ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
PubChem CID 81583
IUPAC Name ethyl 4-methyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)C

Specifications

Color Colorless to Yellow
Density 0.98
Infrared Spectrum Authentic
Refractive Index 1.4255 to 1.4275 (20°C, 589nm)
Linear Formula (CH3)2CHCOCH2CO2C2H5
Quantity 10 g
Beilstein 03, 689
Formula Weight 158.2
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Ethyl isobutyrylacetate, 99%
Show More Show Less
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.