Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Isocyanatoacetate 97.0+%, TCI America™
SDP

Catalog No. I03865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03865G 5 g
I038625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03865G Supplier TCI America Supplier No. I03865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2949-22-6
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00002040
InChI Key DUVOZUPPHBRJJO-UHFFFAOYSA-N
Synonym ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
PubChem CID 76283
IUPAC Name ethyl 2-isocyanatoacetate
SMILES CCOC(=O)CN=C=O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 2206
Formula Weight 129.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isocyanatoacetate
RTECSNumber AI4450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.