Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Isodehydracetate 97%, Thermo Scientific™

Catalog No. AC118630250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118630250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC118630250 Supplier Thermo Scientific Chemicals Supplier No. 118630250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3385-34-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00006643
InChI Key FBPWNVQUVXSXKS-UHFFFAOYSA-N
Synonym ethyl isodehydroacetate, ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate, ethyl isodehydracetate, 5-carbethoxy-4,6-dimethyl-2-pyrone, 2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester, ethyl 4,6-dimethyl-5-coumalate, 2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester, ethylisodehydroacetate, ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate, acmc-20am57
PubChem CID 76918
IUPAC Name ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate
SMILES CCOC(=O)C1=C(C)OC(=O)C=C1C

Specifications

Color Colorless to Yellow
Density 1.16
Infrared Spectrum Authentic
Refractive Index 1.5135 to 1.5165 (20°C, 589nm)
Quantity 25 g
Beilstein 18, 410
Formula Weight 196.2
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Ethyl isodehydracetate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.