Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isodehydroacetate, 95%, Thermo Scientific™

Catalog No. AC373900250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC373900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC373900250 Supplier Thermo Scientific Chemicals Supplier No. 373900250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3385-34-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00006643
InChI Key FBPWNVQUVXSXKS-UHFFFAOYSA-N
Synonym ethyl isodehydroacetate, ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate, ethyl isodehydracetate, 5-carbethoxy-4,6-dimethyl-2-pyrone, 2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester, ethyl 4,6-dimethyl-5-coumalate, 2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester, ethylisodehydroacetate, ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate, acmc-20am57
PubChem CID 76918
IUPAC Name ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
SMILES CCOC(=O)C1=C(C)OC(=O)C=C1C

Specifications

Melting Point 17°C to 18°C
Density 1.1670g/mL
Boiling Point 290°C to 295°C
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5135 to 1.5165
Quantity 25 g
Beilstein 18,41
Specific Gravity 1.167
Solubility Information Solubility in water: insoluble
Formula Weight 196.2
Percent Purity 95%
Chemical Name or Material Ethyl isodehydroacetate
Show More Show Less
EINECSNumber 222-195-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.