Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl isothiocyanatoacetate

Catalog No. 2796925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-969-25G 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 27-969-25G Supplier Sigma Aldrich Supplier No. 2796925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl isothiocyanatoacetate

Specifications

CAS 24066-82-8
Density 1.171 g/mL (at 25°C (literature))
Boiling Point 104°C to 106°C (7 mmHg)
Molecular Formula C5H7NO2S
Refractive Index n20/D 1.502 (literature)
Linear Formula SCNCH2CO2C2H5
MDL Number MFCD00060677
Quantity 10 mg
Molecular Weight (g/mol) 145.18
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.