Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Isothiocyanatoacetate 98.0+%, TCI America™
SDP

Catalog No. I05585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05585G 5 g
I05581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05585G Supplier TCI America Supplier No. I05585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24066-82-8
Molecular Formula C5H7NO2S
Molecular Weight (g/mol) 145.176
MDL Number MFCD00060677
InChI Key IYPSSPPKMLXXRN-UHFFFAOYSA-N
Synonym Isothiocyanatoacetic Acid Ethyl Ester, Ethoxycarbonylmethyl Isothiocyanate
PubChem CID 32202
IUPAC Name ethyl 2-isothiocyanatoacetate
SMILES CCOC(=O)CN=C=S

Specifications

Boiling Point 215°C
Quantity 5 g
UN Number 2810
Formula Weight 145.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isothiocyanatoacetate
RTECSNumber AI5020000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.