Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl L-Lactate, 98%, Spectrum™ Chemical
SDP

Catalog No. 18610434 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
98%
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-434 200 L 98%
18-610-432 100 mL Amber Glass Bottle
18-610-436 500 mL 98%
18-610-435 4 L 98%
18-610-433 20 L 98%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-610-434 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1187200LTBL
Only null left
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 97-64-3

Specifications

Packaging 98%
CAS Max % 1
Odor Amber Glass Bottle
Quantity 200 L
Percent Purity 98%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.