Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl L-lactate, 99%

Catalog No. AAA109000C
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA109000C 10,000 g
AAA1090036 500 g
AAA109000E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA109000C Supplier Thermo Scientific Chemicals Supplier No. A109000C

CAS: 687-47-8 | C5H10O3 | 118.132 g/mol

Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

Solubility
Miscible with water and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00065359
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O

Specifications

Melting Point -25°C
Boiling Point 152°C to 155°C
Flash Point 46°C (114°F)
Assay Percent Range 99%
Quantity 10,000 g
UN Number UN1192
Beilstein 1720839
Merck Index 14,3817
Optical Rotation −11° (neat)
Formula Weight 118.13
Percent Purity 99%
Chemical Name or Material Ethyl L-lactate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H318-H226-H335
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL LACTATE
RTECSNumber OD5070000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.