Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl L-methioninate hydrochloride | 2899-36-7 | 98.0% | 213.73 g/mol | C7H16ClNO2S | 10 G
SDP

Catalog No. 5000209945
Encompass_Preferred

Ethyl L-methioninate hydrochloride is the ethyl ester hydrochloride salt of the amino acid L-methionine, used as an amino acid derivative reagent in research and synthesis applications.

  • Amino acid derivative suitable for biochemical research and synthesis.
  • CAS number 2899-36-7.
  • Molecular formula C7H16ClNO2S and molecular weight 213.73 g/mol.
  • Typical laboratory purity 98.0% and available in small laboratory pack sizes.

Catalog No. 50-002-09945 Supplier Medchemexpress LLC Supplier No. HYW05221210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl L-methioninate hydrochloride is the ethyl ester hydrochloride salt of the amino acid L-methionine, used as an amino acid derivative reagent in research and synthesis applications.

  • Amino acid derivative suitable for biochemical research and synthesis.
  • CAS number 2899-36-7.
  • Molecular formula C7H16ClNO2S and molecular weight 213.73 g/mol.
  • Typical laboratory purity 98.0% and available in small laboratory pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.