Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl L-methioninate hydrochloride | 2899-36-7 | MFCD00012508 | 98.0% | 213.73 g/mol | C7H16ClNO2S | 5 G
SDP

Catalog No. 5000209956
Encompass_Preferred

Ethyl L-methioninate hydrochloride is the hydrochloride salt of the ethyl ester of L-methionine. It is an amino acid derivative used as a research reagent and synthetic intermediate in biochemical and medicinal chemistry applications.

  • High purity: 98.0%.
  • Molecular formula: C7H16ClNO2S.
  • Molecular weight: 213.73 g/mol.
  • Available in laboratory pack sizes, such as 5 g, 10 g, and 50 g.
  • Provided as a crystalline solid suitable for synthesis and analytical use.

Catalog No. 50-002-09956 Supplier Medchemexpress LLC Supplier No. HYW0522125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl L-methioninate hydrochloride is the hydrochloride salt of the ethyl ester of L-methionine. It is an amino acid derivative used as a research reagent and synthetic intermediate in biochemical and medicinal chemistry applications.

  • High purity: 98.0%.
  • Molecular formula: C7H16ClNO2S.
  • Molecular weight: 213.73 g/mol.
  • Available in laboratory pack sizes, such as 5 g, 10 g, and 50 g.
  • Provided as a crystalline solid suitable for synthesis and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.