Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl lactate natural 98 f1KG
Catalog No. 502899793
Encompass
Ethyl lactate is a major volatile flavor compound identified in Chinese Luzhou-flavor type liquor.
Catalog No. 50-289-9793 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2440151KGK
Ethyl lactate is a major volatile flavor compound identified in Chinese Luzhou-flavor type liquor.