Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl Laurate | 106-33-2 | 99.88% | 228.38 | 10 G
SDP

Catalog No. 5000451548
Encompass_Preferred

Ethyl Laurate is an organic compound commonly used as a flavoring and food additive. It can be used to make fruit, mint, and sweet flavors, and is widely used in some food processing and cosmetic production. Additionally, it is utilized as a reagent and intermediate in some organic synthesis reactions. As a biochemical reagent, Ethyl Laurate can be used as a biological material or organic compound for life science related research.

  • Colorless to light yellow liquid
  • Density: 0.9±0.1 g/cm3
  • Shipping at room temperature in continental US
  • Store pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-51548 Supplier Medchemexpress LLC Supplier No. HYW00968410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl Laurate is an organic compound commonly used as a flavoring and food additive. It can be used to make fruit, mint, and sweet flavors, and is widely used in some food processing and cosmetic production. Additionally, it is utilized as a reagent and intermediate in some organic synthesis reactions. As a biochemical reagent, Ethyl Laurate can be used as a biological material or organic compound for life science related research.

  • Colorless to light yellow liquid
  • Density: 0.9±0.1 g/cm3
  • Shipping at room temperature in continental US
  • Store pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.