Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl levulinate

Catalog No. 122629500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-262-9500G 500 g
12-262-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-262-9500G Supplier Sigma Aldrich Supplier No. 122629500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl levulinate is diluent for biodiesel fuels with high saturated fatty acid content.Ethyl levulinate was used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts.

Specifications

CAS 539-88-8
Density 1.016 g/mL (at 25°C (literature))
Boiling Point 93°C to 94°C (18 mmHg)
Molecular Formula C7H12O3
Refractive Index n20/D 1.422 (literature)
Linear Formula CH3COCH2CH2COOC2H5
MDL Number MFCD00009209
Quantity 500 g
Synonym Ethyl 4-oxopentanoate
Molecular Weight (g/mol) 144.17
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.