Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Lignocerate 98.0+%, TCI America™
SDP

Catalog No. L01115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L01115G Supplier TCI America Supplier No. L01115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24634-95-5
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.70
MDL Number MFCD00048673
InChI Key AKXFYSSXNQQBNT-UHFFFAOYSA-N
Synonym Lignoceric Acid Ethyl Ester, Ethyl Tetracosanoate, Tetracosanoic Acid Ethyl Ester
PubChem CID 141135
IUPAC Name ethyl tetracosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 310°C
Quantity 5 g
Formula Weight 396.70
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Lignocerate
EINECSNumber (2)-0798
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.