Learn More
Medchemexpress LLC Ethyl linoleate | 544-35-4 | 99.7% | 308.50 | 1 ML
SDP
Catalog No. 5000454546
Encompass_Preferred
Ethyl linoleate is an orally active unsaturated fatty acid. It inhibits the Akt/GSK3β/β-catenin signaling pathway and the activation of NF-κB. It promotes compound absorption and significantly influences atherosclerosis. It induces heme oxygenase-1 and inhibits tyrosinase, exhibiting whitening and anti-inflammatory effects. It is used in the research of inflammatory diseases and can be used in cosmetics.
  • Inhibits Akt/GSK3β/β-catenin signaling pathway
  • Inhibits activation of NF-κB
  • Induces heme oxygenase-1
  • Inhibits tyrosinase, exhibiting whitening and anti-inflammatory effects
  • Promotes compound absorption
  • Significantly influences atherosclerosis
  • Used in research of inflammatory diseases
  • Can be used in cosmetics
Catalog No. 50-004-54546 Supplier Medchemexpress LLC Supplier No. HYW01381210MM/1ML
Ethyl linoleate is an orally active unsaturated fatty acid. It inhibits the Akt/GSK3β/β-catenin signaling pathway and the activation of NF-κB. It promotes compound absorption and significantly influences atherosclerosis. It induces heme oxygenase-1 and inhibits tyrosinase, exhibiting whitening and anti-inflammatory effects. It is used in the research of inflammatory diseases and can be used in cosmetics.
  • Inhibits Akt/GSK3β/β-catenin signaling pathway
  • Inhibits activation of NF-κB
  • Induces heme oxygenase-1
  • Inhibits tyrosinase, exhibiting whitening and anti-inflammatory effects
  • Promotes compound absorption
  • Significantly influences atherosclerosis
  • Used in research of inflammatory diseases
  • Can be used in cosmetics