Learn More
Medchemexpress LLC Ethyl linoleate (Linoleic Acid ethyl ester) | 544-35-4 | 99.7% | 308.50 | 1 G
SDP
Catalog No. 5000454547
Encompass_Preferred
Ethyl linoleate (Linoleic Acid ethyl ester) is an orally active unsaturated fatty acid. It inhibits the Akt/GSK3β/β-catenin signaling pathway and the activation of NF-κB. This compound induces heme oxygenase-1 and inhibits tyrosinase, leading to whitening and anti-inflammatory effects. Ethyl linoleate also promotes compound absorption and significantly influences atherosclerosis. It is used in the research of inflammatory diseases and can be utilized in cosmetics.
  • Orally active unsaturated fatty acid.
  • Inhibits the Akt/GSK3β/β-catenin signaling pathway and NF-κB activation.
  • Induces heme oxygenase-1.
  • Inhibits tyrosinase.
  • Possesses whitening and anti-inflammatory effects.
  • Promotes compound absorption.
  • Has a significant influence on atherosclerosis.
  • Used in research of inflammatory diseases.
  • Can be used in cosmetics.
Catalog No. 50-004-54547 Supplier Medchemexpress LLC Supplier No. HYW0138121G
Ethyl linoleate (Linoleic Acid ethyl ester) is an orally active unsaturated fatty acid. It inhibits the Akt/GSK3β/β-catenin signaling pathway and the activation of NF-κB. This compound induces heme oxygenase-1 and inhibits tyrosinase, leading to whitening and anti-inflammatory effects. Ethyl linoleate also promotes compound absorption and significantly influences atherosclerosis. It is used in the research of inflammatory diseases and can be utilized in cosmetics.
  • Orally active unsaturated fatty acid.
  • Inhibits the Akt/GSK3β/β-catenin signaling pathway and NF-κB activation.
  • Induces heme oxygenase-1.
  • Inhibits tyrosinase.
  • Possesses whitening and anti-inflammatory effects.
  • Promotes compound absorption.
  • Has a significant influence on atherosclerosis.
  • Used in research of inflammatory diseases.
  • Can be used in cosmetics.