Learn More
Medchemexpress LLC Ethyl linoleate (Linoleic acid ethyl ester) | 544-35-4 | MFCD00009575 | 99.7% | 308.50 | 500 MG
SDP
Catalog No. 5000454557
Encompass_Preferred
Ethyl linoleate is an orally active unsaturated fatty acid, also known as Linoleic Acid ethyl ester. It functions by inhibiting the Akt/GSK3β/β-catenin signaling pathway and the activation of NF-κB. This compound induces heme oxygenase-1 and inhibits tyrosinase, contributing to its whitening and anti-inflammatory effects. It also promotes compound absorption and significantly influences atherosclerosis. Ethyl linoleate is utilized in inflammatory disease research and has applications in cosmetics.
  • Inhibits Akt/GSK3β/β-catenin signaling pathway.
  • Inhibits activation of NF-κB.
  • Induces heme oxygenase-1.
  • Inhibits tyrosinase, contributing to whitening and anti-inflammatory effects.
  • Promotes compound absorption.
  • Significantly influences atherosclerosis.
  • Utilized in inflammatory disease research.
  • Applications in cosmetics.
Catalog No. 50-004-54557 Supplier Medchemexpress LLC Supplier No. HYW013812500MG
Ethyl linoleate is an orally active unsaturated fatty acid, also known as Linoleic Acid ethyl ester. It functions by inhibiting the Akt/GSK3β/β-catenin signaling pathway and the activation of NF-κB. This compound induces heme oxygenase-1 and inhibits tyrosinase, contributing to its whitening and anti-inflammatory effects. It also promotes compound absorption and significantly influences atherosclerosis. Ethyl linoleate is utilized in inflammatory disease research and has applications in cosmetics.
  • Inhibits Akt/GSK3β/β-catenin signaling pathway.
  • Inhibits activation of NF-κB.
  • Induces heme oxygenase-1.
  • Inhibits tyrosinase, contributing to whitening and anti-inflammatory effects.
  • Promotes compound absorption.
  • Significantly influences atherosclerosis.
  • Utilized in inflammatory disease research.
  • Applications in cosmetics.