Learn More
Medchemexpress LLC Ethyl linolenate | 1191-41-9 | 306.48 | 50 MG
SDP
Catalog No. 5000416359
Encompass_Preferred
Ethyl linolenate is a fatty acid ethyl ester (FAEE) that actively inhibits the cellular production of melanin with an IC50 of 70 μM. It significantly blocks Forskolin-induced melanogenesis and inhibits tyrosinase activity.
  • Actively inhibits cellular production of melanin
  • Blocks Forskolin-induced melanogenesis
  • Inhibits tyrosinase activity
  • Demonstrates anti-melanogenesis effects
Catalog No. 50-004-16359 Supplier Medchemexpress LLC Supplier No. HYN207350MG
Ethyl linolenate is a fatty acid ethyl ester (FAEE) that actively inhibits the cellular production of melanin with an IC50 of 70 μM. It significantly blocks Forskolin-induced melanogenesis and inhibits tyrosinase activity.
  • Actively inhibits cellular production of melanin
  • Blocks Forskolin-induced melanogenesis
  • Inhibits tyrosinase activity
  • Demonstrates anti-melanogenesis effects