Learn More
Medchemexpress LLC Ethyl linolenate | 1191-41-9 | MFCD07371452 | 306.48 | 100 MG
SDP
Catalog No. 5000416170
Encompass_Preferred
Ethyl linolenate is a fatty acid ethyl ester (FAEE) that actively inhibits the cellular production of melanin with an IC50 of 70 μM. It exhibits anti-melanogenesis effects.
  • Fatty acid ethyl ester (FAEE)
  • Inhibits cellular melanin production
  • Significantly blocks Forskolin-induced melanogenesis
  • Inhibits tyrosinase activity
Catalog No. 50-004-16170 Supplier Medchemexpress LLC Supplier No. HYN2073100MG
Ethyl linolenate is a fatty acid ethyl ester (FAEE) that actively inhibits the cellular production of melanin with an IC50 of 70 μM. It exhibits anti-melanogenesis effects.
  • Fatty acid ethyl ester (FAEE)
  • Inhibits cellular melanin production
  • Significantly blocks Forskolin-induced melanogenesis
  • Inhibits tyrosinase activity