Learn More
Medchemexpress LLC Ethyl maltol (2-Ethyl-3-hydroxy-4H-pyran-4-one) | 4940-11-8 | 100.0% | 140.14 | 1 ML
SDP
Catalog No. 5000454033
Encompass_Preferred
Ethyl maltol (2-Ethyl-3-hydroxy-4H-pyran-4-one) is an orally active and important food additive and flavor enhancer. Ethyl maltol is less toxic to rats and dogs. Ethyl maltol can enhance copper-mediated cytotoxicity and induce apoptosis in lung epithelial cells.
  • Orally active.
  • Important food additive.
  • Flavor enhancer.
  • Less toxic to rats and dogs.
  • Can enhance copper-mediated cytotoxicity.
  • Can induce apoptosis in lung epithelial cells.
  • Induces cell apoptosis and leads to oxidative stress and DNA damage in copper treated lung epithelial cells.
  • Increases pATM phosphorylation level.
  • Increases the expression of ferritin light chain and hemeoxygenase.
  • Safe in rats and dogs.
  • Toxicity is less than the homolog maltol.
Catalog No. 50-004-54033 Supplier Medchemexpress LLC Supplier No. HYW01032010MM/1ML
Ethyl maltol (2-Ethyl-3-hydroxy-4H-pyran-4-one) is an orally active and important food additive and flavor enhancer. Ethyl maltol is less toxic to rats and dogs. Ethyl maltol can enhance copper-mediated cytotoxicity and induce apoptosis in lung epithelial cells.
  • Orally active.
  • Important food additive.
  • Flavor enhancer.
  • Less toxic to rats and dogs.
  • Can enhance copper-mediated cytotoxicity.
  • Can induce apoptosis in lung epithelial cells.
  • Induces cell apoptosis and leads to oxidative stress and DNA damage in copper treated lung epithelial cells.
  • Increases pATM phosphorylation level.
  • Increases the expression of ferritin light chain and hemeoxygenase.
  • Safe in rats and dogs.
  • Toxicity is less than the homolog maltol.