Learn More
Medchemexpress LLC Ethyl maltol-d5 | 4940-11-8 | 99.0% | 145.17 | 1 MG
SDP
Catalog No. 5000453757
Encompass_Preferred
Ethyl maltol-d5 is the deuterium labeled Ethyl maltol. Ethyl maltol is an orally active and important food additive and flavor enhancer. It is less toxic to rats and dogs and can enhance copper-mediated cytotoxicity and induce apoptosis in lung epithelial cells. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53757 Supplier Medchemexpress LLC Supplier No. HYW010320S1MG
Ethyl maltol-d5 is the deuterium labeled Ethyl maltol. Ethyl maltol is an orally active and important food additive and flavor enhancer. It is less toxic to rats and dogs and can enhance copper-mediated cytotoxicity and induce apoptosis in lung epithelial cells. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect the pharmacokinetic and metabolic profiles of drugs.