Learn More
Medchemexpress LLC Ethyl maltol (Standard) | 4940-11-8 | 99.98% | 140.14 | 50 MG
SDP
Catalog No. 5000451856
Encompass_Preferred
Ethyl maltol (Standard) is the analytical standard of Ethyl maltol, intended for research and analytical applications. It is an orally active and important food additive and flavor enhancer. It is less toxic to rats and dogs and can enhance copper-mediated cytotoxicity and induce apoptosis in lung epithelial cells.
  • Analytical standard of Ethyl maltol
  • Intended for research and analytical applications
  • Orally active
  • Important food additive and flavor enhancer
  • Less toxic to rats and dogs
  • Can enhance copper-mediated cytotoxicity
  • Induces apoptosis in lung epithelial cells
Catalog No. 50-004-51856 Supplier Medchemexpress LLC Supplier No. HYW010320R50MG
Ethyl maltol (Standard) is the analytical standard of Ethyl maltol, intended for research and analytical applications. It is an orally active and important food additive and flavor enhancer. It is less toxic to rats and dogs and can enhance copper-mediated cytotoxicity and induce apoptosis in lung epithelial cells.
  • Analytical standard of Ethyl maltol
  • Intended for research and analytical applications
  • Orally active
  • Important food additive and flavor enhancer
  • Less toxic to rats and dogs
  • Can enhance copper-mediated cytotoxicity
  • Induces apoptosis in lung epithelial cells