Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl mandelate | 774-40-3 | MFCD00004494 | 99.7% | 180.20 g·mol⁻1 | C10H12O3 | 50 G
SDP

Catalog No. 5000301238
Encompass_Preferred

Ethyl mandelate is the ethyl ester of mandelic acid commonly used as an intermediate in pharmaceutical and organic synthesis. It is supplied for research and synthetic chemistry applications with documentation available for quality and safety.

  • Used as a pharmaceutical intermediate and synthetic building block.
  • Purity suitable for research applications (99.7%).
  • Molecular weight 180.20 g·mol⁻1 and formula C10H12O3.
  • Available in multiple pack sizes for laboratory use.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-003-01238 Supplier Medchemexpress LLC Supplier No. HYW02703850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ethyl mandelate is the ethyl ester of mandelic acid commonly used as an intermediate in pharmaceutical and organic synthesis. It is supplied for research and synthetic chemistry applications with documentation available for quality and safety.

  • Used as a pharmaceutical intermediate and synthetic building block.
  • Purity suitable for research applications (99.7%).
  • Molecular weight 180.20 g·mol⁻1 and formula C10H12O3.
  • Available in multiple pack sizes for laboratory use.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.