Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Ethyl mandelate, 97%

Catalog No. AAA1741222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1741222 100 g
AAA1741214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1741222 Supplier Thermo Scientific Chemicals Supplier No. A1741222
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 774-40-3 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-40-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00004494
InChI Key SAXHIDRUJXPDOD-UHFFFAOYSA-N
Synonym ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
PubChem CID 13050
ChEBI CHEBI:38750
IUPAC Name ethyl 2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Density 1.115
Boiling Point 253°C to 255°C
Flash Point >110°C (230°F)
Quantity 100 g
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material (+/-)-Ethyl mandelate
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 212-263-0
RTECSNumber OO7397000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.