Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl methacrylate

Catalog No. 234893100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-489-3100ML 100 mL
23-489-31L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-489-3100ML Supplier Sigma Aldrich Supplier No. 234893100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl methacrylate is a readily polymerizable monomer used for certain types of acrylic resins. The monomethyl ether hydroxyl quinone present in it is an inhibitor that prevents polymerization.Ethyl methacrylate (EMA) may be used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It may be used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.

Specifications

CAS 97-63-2
Density 0.917 g/mL (at 25°C)
Boiling Point 118°C to 119°C
Molecular Formula C6H10O2
Refractive Index n20/D 1.413 (literature)
Linear Formula CH2=C(CH3)COOC2H5
MDL Number MFCD00009161
Quantity 100 mL
Synonym 2-Methyl-2-propenoic acid
Molecular Weight (g/mol) 114.14
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.