Learn More
Medchemexpress LLC Ethyl methanesulfonate | 62-50-0 | MFCD00007559 | 100.0% | 124.15 g·mol^-1 | C3H8O3S | 5 G
SDP
Catalog No. 5000299918
Encompass_Preferred
Ethyl methanesulfonate (EMS) is a small-molecule alkylating agent used primarily as a chemical mutagen in genetic toxicology and mutagenesis research. It alkylates DNA to induce point mutations and other genetic alterations, and is handled as a hazardous mutagen and suspected carcinogen; appropriate containment and PPE are required when in use.
  • Induces point mutations via DNA alkylation.
  • Widely used for mutagenesis and genetic toxicology studies.
  • Available in high-purity grades suitable for research.
  • Small molecular weight compound (~124.15 g·mol^-1).
  • Requires strict safety precautions due to mutagenic and carcinogenic properties.
Catalog No. 50-002-99918 Supplier Medchemexpress LLC Supplier No. HYW0158545G
Ethyl methanesulfonate (EMS) is a small-molecule alkylating agent used primarily as a chemical mutagen in genetic toxicology and mutagenesis research. It alkylates DNA to induce point mutations and other genetic alterations, and is handled as a hazardous mutagen and suspected carcinogen; appropriate containment and PPE are required when in use.
  • Induces point mutations via DNA alkylation.
  • Widely used for mutagenesis and genetic toxicology studies.
  • Available in high-purity grades suitable for research.
  • Small molecular weight compound (~124.15 g·mol^-1).
  • Requires strict safety precautions due to mutagenic and carcinogenic properties.