Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl methanesulfonate | 62-50-0 | MFCD00007559 | 98.3% | 124.15 g/mol | C3H8O3S | 1 G
SDP

Catalog No. 5000300540
Encompass_Preferred

Ethyl methanesulfonate is a small-molecule alkylating agent used in genetic toxicology and mutagenesis research to induce point mutations and study DNA damage responses. Supplied as a liquid reagent for research use only, it requires careful handling and adherence to safety data sheet recommendations due to its mutagenic properties.

  • Alkylating mutagen for inducing point mutations.
  • Liquid reagent, colorless to light yellow.
  • Typical purity 98.33% (specific batches up to 99.96%).
  • Molecular weight 124.15 g/mol; formula C3H8O3S.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • SDS and handling information available.

Catalog No. 50-003-00540 Supplier Medchemexpress LLC Supplier No. HYW0158541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl methanesulfonate is a small-molecule alkylating agent used in genetic toxicology and mutagenesis research to induce point mutations and study DNA damage responses. Supplied as a liquid reagent for research use only, it requires careful handling and adherence to safety data sheet recommendations due to its mutagenic properties.

  • Alkylating mutagen for inducing point mutations.
  • Liquid reagent, colorless to light yellow.
  • Typical purity 98.33% (specific batches up to 99.96%).
  • Molecular weight 124.15 g/mol; formula C3H8O3S.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • SDS and handling information available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.