Learn More
Medchemexpress LLC Ethyl methanesulfonate | 62-50-0 | MFCD00007559 | 98.3% | 124.15 g/mol | C3H8O3S | 1 ML
SDP
Catalog No. 5000303762
Encompass_Preferred
Ethyl methanesulfonate (EMS) is a small-molecule alkylating reagent supplied as a neat liquid or a ready-to-use 10 mM solution in DMSO. It is commonly used in biological and genetic research to induce ethylation of nucleic acids and study mutagenesis. Handle with appropriate safety controls; for research use only.
  • Ready-to-use 10 mM solution in DMSO for direct applications.
  • Available in liquid and bulk quantities to suit experimental scale.
  • High purity suitable for analytical and research purposes.
  • Effective alkylating agent for mutagenesis and DNA modification studies.
  • Provided with documentation to support handling and safety.
Catalog No. 50-003-03762 Supplier Medchemexpress LLC Supplier No. HYW01585410MM/1ML
Ethyl methanesulfonate (EMS) is a small-molecule alkylating reagent supplied as a neat liquid or a ready-to-use 10 mM solution in DMSO. It is commonly used in biological and genetic research to induce ethylation of nucleic acids and study mutagenesis. Handle with appropriate safety controls; for research use only.
  • Ready-to-use 10 mM solution in DMSO for direct applications.
  • Available in liquid and bulk quantities to suit experimental scale.
  • High purity suitable for analytical and research purposes.
  • Effective alkylating agent for mutagenesis and DNA modification studies.
  • Provided with documentation to support handling and safety.