Learn More
Medchemexpress LLC Ethyl methanesulfonate-d5 | 1219795-44-4 | 99.5% | 129.19 | C3H3D5O3S | 5 MG
SDP
Catalog No. 5000300256
Encompass_Preferred
Ethyl methanesulfonate-d5 is a deuterium-labeled analogue of ethyl methanesulfonate intended for use as an internal standard and tracer in analytical and drug metabolism studies. The product is provided as a high-purity isotopically labeled liquid with accompanying documentation for laboratory use and recommended storage conditions.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity for accurate and reproducible results.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Available in small vials suitable for analytical applications.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-003-00256 Supplier Medchemexpress LLC Supplier No. HYW015854S5MG
Ethyl methanesulfonate-d5 is a deuterium-labeled analogue of ethyl methanesulfonate intended for use as an internal standard and tracer in analytical and drug metabolism studies. The product is provided as a high-purity isotopically labeled liquid with accompanying documentation for laboratory use and recommended storage conditions.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity for accurate and reproducible results.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Available in small vials suitable for analytical applications.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Stable under recommended storage conditions for long-term use.