Learn More
Medchemexpress LLC Ethyl methanesulfonate-d5 | 1219795-44-4 | 99.5% | 129.19 g/mol | C3H3D5O3S | 10 MG
SDP
Catalog No. 5000300255
Encompass_Preferred
Ethyl methanesulfonate-d5 is the deuterium-labeled analogue of ethyl methanesulfonate used as a stable-isotope reference standard for analytical and research applications. Supplied as a colorless to light yellow liquid with high isotopic and chemical purity, it is intended for use in mass spectrometry, method development, and quantitative studies.
  • High isotopic enrichment and chemical purity (≈99.5%).
  • Optimized for mass spectrometry and quantitative analysis.
  • Useful as an internal standard for method development and validation.
  • Available in small research pack sizes suitable for laboratory use.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality and handling information.
Catalog No. 50-003-00255 Supplier Medchemexpress LLC Supplier No. HYW015854S10MG
Ethyl methanesulfonate-d5 is the deuterium-labeled analogue of ethyl methanesulfonate used as a stable-isotope reference standard for analytical and research applications. Supplied as a colorless to light yellow liquid with high isotopic and chemical purity, it is intended for use in mass spectrometry, method development, and quantitative studies.
  • High isotopic enrichment and chemical purity (≈99.5%).
  • Optimized for mass spectrometry and quantitative analysis.
  • Useful as an internal standard for method development and validation.
  • Available in small research pack sizes suitable for laboratory use.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality and handling information.