Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl methyl carbonate

Catalog No. 75493550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-493-550ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-493-550ML Supplier Sigma Aldrich Supplier No. 75493550ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.EMC is majorly utilized as a co-solvent in the nonaqueous electrolyte. It enhances the energy density and discharge capacity of lithium-ion batteries.

Specifications

CAS 623-53-0
Density 1.006 g/mL (at 25°C)
Molecular Formula C4H8O3
Refractive Index n20/D 1.378
Linear Formula C4H8O3
MDL Number MFCD00191398
Quantity 50 mL
Synonym Carbonic acid ethyl methyl ester; EMC; Methyl ethyl carbonate
Molecular Weight (g/mol) 104.1
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.