Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl methylcarbamate, 99%, Thermo Scientific™

Catalog No. AC410265000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC410265000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC410265000 Supplier Thermo Scientific Chemicals Supplier No. 410265000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-40-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00041924
InChI Key SURZCVYFPAXNGN-UHFFFAOYSA-N
Synonym ethyl methylcarbamate, n-methylurethane, n-methylurethan, ethyl-n-methylcarbamate, methylcarbamic acid, ethyl ester, n-methyl urethan, carbamic acid, methyl-, ethyl ester, methylurethane van, unii-1w34gcf5cs, ccris 2510
PubChem CID 7752
IUPAC Name ethyl N-methylcarbamate
SMILES CCOC(=O)NC

Specifications

Boiling Point 170.0°C
Refractive Index 1.4180 to 1.4200
Linear Formula CH3NHCOOC2H5
Solubility Information Solubility in water: 6900g/L (16°C). Other solubilities: soluble in alcohol
Density 1.0100g/mL
Quantity 500 g
Flash Point 61°C
Formula Weight 103.12
Specific Gravity 1.01
Chemical Name or Material Ethyl methylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.