Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl N-(2,2-diphenylacetyl)carbamate | 302841-86-7 | MFCD20270619 | 99.9% | 283.32 g/mol | C17H17NO3 | 1 ML
SDP

Catalog No. 5000240577
Encompass_Preferred

Ro 01-6128 is a small-molecule positive allosteric modulator of metabotropic glutamate receptor 1 (mGluR1) supplied for research applications. It is provided as a 10 mM solution in DMSO or as solid quantities, and has documented solubility, storage, and purity data to support in vitro and in vivo studies.

  • Positive allosteric modulator of mGluR1 for receptor pharmacology studies.
  • High purity reported at 99.87%.
  • Molecular weight 283.32 g/mol; formula C17H17NO3.
  • Available as 10 mM solution in DMSO and as solid sizes for formulation.
  • Soluble in DMSO (≈100 mg/mL); may require ultrasonic agitation.
  • Storage recommendations include powder at -20°C and prepared solutions at -80°C for extended stability.

Catalog No. 50-002-40577 Supplier Medchemexpress LLC Supplier No. HY10750710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 01-6128 is a small-molecule positive allosteric modulator of metabotropic glutamate receptor 1 (mGluR1) supplied for research applications. It is provided as a 10 mM solution in DMSO or as solid quantities, and has documented solubility, storage, and purity data to support in vitro and in vivo studies.

  • Positive allosteric modulator of mGluR1 for receptor pharmacology studies.
  • High purity reported at 99.87%.
  • Molecular weight 283.32 g/mol; formula C17H17NO3.
  • Available as 10 mM solution in DMSO and as solid sizes for formulation.
  • Soluble in DMSO (≈100 mg/mL); may require ultrasonic agitation.
  • Storage recommendations include powder at -20°C and prepared solutions at -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.