Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl n-Valerate, MP Biomedicals

Catalog No. ICN202835 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202835 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202835 Supplier MP Biomedicals, Inc Supplier No. 0520283580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 539-82-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
InChI Key ICMAFTSLXCXHRK-UHFFFAOYSA-N
Synonym ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
PubChem CID 10882
IUPAC Name ethyl pentanoate
SMILES CCCCC(=O)OCC

Specifications

Quantity 100 g
Formula Weight 130.18
Chemical Name or Material Ethyl N-Valerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.